missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzenethiol 97.0+%, TCI America™

Catalog No. T25191G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25191G 1 g
T25195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25191G Supplier TCI America Supplier No. T25191G

Chemical Identifiers

CAS 13333-97-6
Molecular Formula C7H5F3S
Molecular Weight (g/mol) 178.172
MDL Number MFCD00068896
InChI Key XVGQHRKNXSUPEF-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzenethiol, 2-trifluoromethyl thiophenol, 2-trifluoromethylthiophenol, 2-trifluoromethyl benzene-1-thiol, 2-mercaptobenzotrifluoride, pubchem6860, acmc-209brx, trifluoromethyl thiophenol, 2trifluoromethylbenzenethiol
PubChem CID 2777889
IUPAC Name 2-(trifluoromethyl)benzenethiol
SMILES C1=CC=C(C(=C1)C(F)(F)F)S

Specifications

Color Yellow
Boiling Point 64°C
Quantity 1 g
UN Number 2810
Formula Weight 178.17
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethyl)benzenethiol