missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Trifluoromethyl)benzenesulfonyl Chloride 98.0+%, TCI America™
$55.84 - $219.48
Chemical Identifiers
| CAS | 776-04-5 |
|---|---|
| Molecular Formula | C7H4ClF3O2S |
| Molecular Weight (g/mol) | 244.612 |
| InChI Key | ZIZGWNOAHUCACM-UHFFFAOYSA-N |
| Synonym | 2-trifluoromethyl benzenesulfonyl chloride, 2-trifluoromethyl benzene-1-sulfonyl chloride, 2-trifluoromethylbenzenesulphonyl chloride, 2-trifluoromethyl benzenesulphonyl chloride, o-trifluoromethyl benzenesulfonyl chloride, o-trifluoromethylbenzenesulfonyl chloride, 2-trifluoromethyl benzene sulfonyl chloride, benzenesulfonyl chloride, 2-trifluoromethyl, alpha,alpha,alpha-trifluorotoluene-2-sulfonyl chloride |
| PubChem CID | 136616 |
| IUPAC Name | 2-(trifluoromethyl)benzenesulfonyl chloride |
| SMILES | C1=CC=C(C(=C1)C(F)(F)F)S(=O)(=O)Cl |
Chemical Identifiers
| 776-04-5 | |
| 244.612 | |
| 2-trifluoromethyl benzenesulfonyl chloride, 2-trifluoromethyl benzene-1-sulfonyl chloride, 2-trifluoromethylbenzenesulphonyl chloride, 2-trifluoromethyl benzenesulphonyl chloride, o-trifluoromethyl benzenesulfonyl chloride, o-trifluoromethylbenzenesulfonyl chloride, 2-trifluoromethyl benzene sulfonyl chloride, benzenesulfonyl chloride, 2-trifluoromethyl, alpha,alpha,alpha-trifluorotoluene-2-sulfonyl chloride | |
| 2-(trifluoromethyl)benzenesulfonyl chloride |
| C7H4ClF3O2S | |
| ZIZGWNOAHUCACM-UHFFFAOYSA-N | |
| 136616 | |
| C1=CC=C(C(=C1)C(F)(F)F)S(=O)(=O)Cl |
Specifications
| 776-04-5 | |
| 126°C | |
| 1 g | |
| 2-trifluoromethyl benzenesulfonyl chloride, 2-trifluoromethyl benzene-1-sulfonyl chloride, 2-trifluoromethylbenzenesulphonyl chloride, 2-trifluoromethyl benzenesulphonyl chloride, o-trifluoromethyl benzenesulfonyl chloride, o-trifluoromethylbenzenesulfonyl chloride, 2-trifluoromethyl benzene sulfonyl chloride, benzenesulfonyl chloride, 2-trifluoromethyl, alpha,alpha,alpha-trifluorotoluene-2-sulfonyl chloride | |
| C1=CC=C(C(=C1)C(F)(F)F)S(=O)(=O)Cl | |
| 244.612 | |
| 244.61 | |
| Liquid |
| Yellow | |
| C7H4ClF3O2S | |
| 3265 | |
| ZIZGWNOAHUCACM-UHFFFAOYSA-N | |
| 2-(trifluoromethyl)benzenesulfonyl chloride | |
| 136616 | |
| ≥98.0% (GC) | |
| 2-(Trifluoromethyl)benzenesulfonyl Chloride |
Safety and Handling
TSCA : No