missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. T18921G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18921G 1 g
T18925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18921G Supplier TCI America Supplier No. T18921G

Chemical Identifiers

CAS 776-04-5
Molecular Formula C7H4ClF3O2S
Molecular Weight (g/mol) 244.612
InChI Key ZIZGWNOAHUCACM-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzenesulfonyl chloride, 2-trifluoromethyl benzene-1-sulfonyl chloride, 2-trifluoromethylbenzenesulphonyl chloride, 2-trifluoromethyl benzenesulphonyl chloride, o-trifluoromethyl benzenesulfonyl chloride, o-trifluoromethylbenzenesulfonyl chloride, 2-trifluoromethyl benzene sulfonyl chloride, benzenesulfonyl chloride, 2-trifluoromethyl, alpha,alpha,alpha-trifluorotoluene-2-sulfonyl chloride
PubChem CID 136616
IUPAC Name 2-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=CC=C(C(=C1)C(F)(F)F)S(=O)(=O)Cl

Specifications

Color Yellow
Boiling Point 126°C
Quantity 1 g
UN Number 3265
Formula Weight 244.61
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethyl)benzenesulfonyl Chloride