Learn More
2-(Trifluoromethyl)benzaldehyde, 98%
CAS: 447-61-0 | C8H5F3O | 174.122 g/mol
$74.51 - $434.26
Chemical Identifiers
| CAS | 447-61-0 |
|---|---|
| Molecular Formula | C8H5F3O |
| Molecular Weight (g/mol) | 174.122 |
| MDL Number | MFCD00003337 |
| InChI Key | ZDVRPKUWYQVVDX-UHFFFAOYSA-N |
| Synonym | 2-trifluoromethyl benzaldehyde, o-trifluoromethyl benzaldehyde, o-trifluoromethylbenzaldehyde, alpha,alpha,alpha-trifluoro-o-tolualdehyde, benzaldehyde, 2-trifluoromethyl, 2-trifluoromethyl-benzaldehyde, ortho-trifluoromethyl-benzaldehyde, otf-bad, trifluoromethylbenzaldehyde |
| PubChem CID | 123067 |
| IUPAC Name | 2-(trifluoromethyl)benzaldehyde |
| SMILES | C1=CC=C(C(=C1)C=O)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2459106
|
Thermo Scientific Chemicals
B2459106 |
5 g |
Each for $74.51
|
|
|||||
|
AAB2459114
|
Thermo Scientific Chemicals
B2459114 |
25 g |
Each for $144.59
|
|
|||||
|
AAB2459122
|
Thermo Scientific Chemicals
B2459122 |
100 g |
Each for $434.26
|
|
|||||
Description
2-(Trifluoromethyl)benzaldehyde has been used in the preparation of ω-(dimethylamino)-alkylethyl-4(-2-(trifluorormethyl)phenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylates, methyl (E)-2-[2-[(E)-2-(2-trifluoromethylphenyl)vinyl]-phenyl]-3-methoxyacrylate via Wittig-Horner reaction. It is a reagent used as electrophilic component in a wide array of reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(Trifluoromethyl)benzaldehyde has been used in the preparation of ω-(dimethylamino)-alkylethyl-4(-2-(trifluorormethyl)phenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylates, methyl (E)-2-[2-[(E)-2-(2-trifluoromethylphenyl)vinyl]-phenyl]-3-methoxyacrylate via Wittig-Horner reaction. It is a reagent used as electrophilic component in a wide array of reactions.
Solubility
Soluble in most organic solvents.
Notes
Air sensitive. Store under dry inert gas. Store away from oxidizing agents, air.
Chemical Identifiers
| 447-61-0 | |
| 174.122 | |
| ZDVRPKUWYQVVDX-UHFFFAOYSA-N | |
| 123067 | |
| C1=CC=C(C(=C1)C=O)C(F)(F)F |
| C8H5F3O | |
| MFCD00003337 | |
| 2-trifluoromethyl benzaldehyde, o-trifluoromethyl benzaldehyde, o-trifluoromethylbenzaldehyde, alpha,alpha,alpha-trifluoro-o-tolualdehyde, benzaldehyde, 2-trifluoromethyl, 2-trifluoromethyl-benzaldehyde, ortho-trifluoromethyl-benzaldehyde, otf-bad, trifluoromethylbenzaldehyde | |
| 2-(trifluoromethyl)benzaldehyde |
Specifications
| 447-61-0 | |
| 1.322 | |
| 61°C (141°F) | |
| C8H5F3O | |
| MFCD00003337 | |
| 2045512 | |
| 2-trifluoromethyl benzaldehyde, o-trifluoromethyl benzaldehyde, o-trifluoromethylbenzaldehyde, alpha,alpha,alpha-trifluoro-o-tolualdehyde, benzaldehyde, 2-trifluoromethyl, 2-trifluoromethyl-benzaldehyde, ortho-trifluoromethyl-benzaldehyde, otf-bad, trifluoromethylbenzaldehyde | |
| ZDVRPKUWYQVVDX-UHFFFAOYSA-N | |
| 2-(trifluoromethyl)benzaldehyde | |
| 123067 | |
| 98% |
| -40°C | |
| 70°C to 71°C (16 mmHg) | |
| Almond-like | |
| 1.466 | |
| 5 g | |
| Air Sensitive | |
| Soluble in most organic solvents. | |
| C1=CC=C(C(=C1)C=O)C(F)(F)F | |
| 174.122 | |
| 174.12 | |
| 2-(Trifluoromethyl)benzaldehyde |
Safety and Handling
GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
EINECSNumber : 207-185-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only