missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzaldehyde, 97%

Catalog No. AC232680250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC232680250 25 g Glass bottle
AC232681000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC232680250 Supplier Thermo Scientific Chemicals Supplier No. 232680250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 447-61-0 | C8H5F3O | 174.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 447-61-0
Molecular Formula C8H5F3O
Molecular Weight (g/mol) 174.12
MDL Number MFCD00003337
InChI Key ZDVRPKUWYQVVDX-UHFFFAOYSA-N
Synonym 2-trifluoromethyl benzaldehyde, o-trifluoromethyl benzaldehyde, o-trifluoromethylbenzaldehyde, alpha,alpha,alpha-trifluoro-o-tolualdehyde, benzaldehyde, 2-trifluoromethyl, 2-trifluoromethyl-benzaldehyde, ortho-trifluoromethyl-benzaldehyde, otf-bad, trifluoromethylbenzaldehyde
PubChem CID 123067
IUPAC Name 2-(trifluoromethyl)benzaldehyde
SMILES C1=CC=C(C(=C1)C=O)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -40.0°C
Color Colorless to Yellow
Density 1.3200g/mL
Boiling Point 70.0°C to 71.0°C (16.0 mmHg)
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4650 to 1.4670
Linear Formula CF3C6H4CHO
Quantity 25 g
Beilstein 07, III, 1013
Specific Gravity 1.32
Formula Weight 174.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethyl)benzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 207-185-9

RUO – Research Use Only