missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzal Chloride, TCI America™

Catalog No. T130525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T130525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T130525G Supplier TCI America Supplier No. T130525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 707-72-2
Molecular Formula C8H5Cl2F3
Molecular Weight (g/mol) 229.02
MDL Number MFCD00077613
InChI Key JIJFXGFHPXLJME-UHFFFAOYSA-N
PubChem CID 606399
IUPAC Name 1-(dichloromethyl)-2-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC=CC=C1C(Cl)Cl

Specifications

Color Colorless
Boiling Point 71°C
Quantity 25 g
UN Number 1760
Formula Weight 229.02
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethyl)benzal Chloride