missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)acrylic Acid 98.0+%, TCI America™

Catalog No. p-7136812
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15125G 5 g
T151225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15125G Supplier TCI America Supplier No. T15125G
Only null left

Chemical Identifiers

CAS 381-98-6
Molecular Formula C4H2F3O2
Molecular Weight (g/mol) 139.05
MDL Number MFCD00042424
InChI Key VLSRKCIBHNJFHA-UHFFFAOYSA-M
Synonym 2-trifluoromethyl acrylic acid, 2-trifluoromethyl propenoic acid, 2-trifluoromethyl prop-2-enoic acid, tfmaa, trifluoromethacrylic acid, trifluoromethylacrylic acid, 2-trifluoromethylacrylic acid, 2-trifluoromethyl-acrylic acid, 2-propenoic acid, 2-trifluoromethyl, pubchem22654
PubChem CID 587694
IUPAC Name 2-(trifluoromethyl)prop-2-enoate
SMILES [O-]C(=O)C(=C)C(F)(F)F

Specifications

Melting Point 52°C
Boiling Point 148°C
Quantity 5 g
UN Number 3261
Formula Weight 140.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Trifluoromethyl)acrylic Acid