accessibility menu, dialog, popup

UserName

CAS RN 381-98-6

IUPAC Name: 2-(trifluoromethyl)prop-2-enoate
Synonyms: 2-(trifluoromethyl)prop-2-enoate
Molecular Weight (g/mol): 139.05
Molecular Formula: C4H2F3O2
InChi Key: VLSRKCIBHNJFHA-UHFFFAOYSA-M
SMILES: [O-]C(=O)C(=C)C(F)(F)F
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)