missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)acrylic acid, 98%

Catalog No. p-7026445
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2244303 1 g
AAB2244306 5 g
AAB2244314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2244303 Supplier Thermo Scientific Chemicals Supplier No. B2244303
Only null left

CAS: 381-98-6 | C4H2F3O2 | 139.05 g/mol

2-(Trifluoromethyl)acrylic acid is used as an acidic functional monomer for molecular imprinting of nicotine. And also used in the conventional functional monomer methacrylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)acrylic acid is used as an acidic functional monomer for molecular imprinting of nicotine. And also used in the conventional functional monomer methacrylic acid.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 381-98-6
Molecular Formula C4H2F3O2
Molecular Weight (g/mol) 139.05
MDL Number MFCD00042424
InChI Key VLSRKCIBHNJFHA-UHFFFAOYSA-M
Synonym 2-trifluoromethyl acrylic acid, 2-trifluoromethyl propenoic acid, 2-trifluoromethyl prop-2-enoic acid, tfmaa, trifluoromethacrylic acid, trifluoromethylacrylic acid, 2-trifluoromethylacrylic acid, 2-trifluoromethyl-acrylic acid, 2-propenoic acid, 2-trifluoromethyl, pubchem22654
PubChem CID 587694
SMILES [O-]C(=O)C(=C)C(F)(F)F

Specifications

Melting Point 49°C to 51°C
Flash Point 82°C (180°F)
Quantity 1 g
UN Number UN3261
Solubility Information Soluble in water.
IUPAC Name 2-(trifluoromethyl)prop-2-enoic acid
Formula Weight 140.06
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)acrylic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only