missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)acrylic acid, 98%

Catalog No. AC313100010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC313100010 1 g Glass bottle
AC313100050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC313100010 Supplier Thermo Scientific Chemicals Supplier No. 313100010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 381-98-6 | C4H2F3O2 | 139.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 381-98-6
Molecular Formula C4H2F3O2
Molecular Weight (g/mol) 139.05
MDL Number MFCD00042424
InChI Key VLSRKCIBHNJFHA-UHFFFAOYSA-M
Synonym 2-trifluoromethyl acrylic acid, 2-trifluoromethyl propenoic acid, 2-trifluoromethyl prop-2-enoic acid, tfmaa, trifluoromethacrylic acid, trifluoromethylacrylic acid, 2-trifluoromethylacrylic acid, 2-trifluoromethyl-acrylic acid, 2-propenoic acid, 2-trifluoromethyl, pubchem22654
PubChem CID 587694
IUPAC Name 2-(trifluoromethyl)prop-2-enoic acid
SMILES [O-]C(=O)C(=C)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 48°C to 52°C
Color Yellow
Boiling Point 86°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2C=C(CF3)CO2H
Quantity 1 g
Formula Weight 140.06
Percent Purity 98%
Physical Form Chunks
Chemical Name or Material 2-(Trifluoromethyl)acrylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only