missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)aniline 98.0+%, TCI America™

Numéro de catalogue. T21455G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T21455G 5 g
T214525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T21455G Fournisseur TCI America Code fournisseur T21455G
Il en reste null

Identifiants chimiques

CAS 1535-75-7
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.126
InChI Key ZFCOUBUSGHLCDT-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy aniline, benzenamine, 2-trifluoromethoxy, 2-trifluoromethoxy-phenylamine, o-trifluoromethoxy aniline, alpha,alpha,alpha-trifluoro-o-anisidine, o-trifluoromethoxyaniline, 2-trifluoromethoxy aminobenzene, 2-trifluoromethoxy phenylamine, 2-trifluoromethoxy phenyl amine
PubChem CID 73754
IUPAC Name 2-(trifluoromethoxy)aniline
SMILES C1=CC=C(C(=C1)N)OC(F)(F)F

Spécifications

Boiling Point 63°C
Quantity 5 g
UN Number 1993
Formula Weight 177.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Trifluoromethoxy)aniline