missing translation for 'onlineSavingsMsg'
Learn More

2-(Tri-n-butylstannyl)thiazole, 96%

Catalog No. p-7033896
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5109003 1 g
AAH5109006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5109003 Supplier Thermo Scientific Chemicals Supplier No. H5109003
Only null left

CAS: 121359-48-6 | C15H29NSSn | 374.174 g/mol

Used as reagent for arylation of thiazole by Stille cross-coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as reagent for arylation of thiazole by Stille cross-coupling.

Solubility
Not miscible or difficult to mix.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 121359-48-6
Molecular Formula C15H29NSSn
Molecular Weight (g/mol) 374.174
MDL Number MFCD01319057
InChI Key WUOFQGMXQCSPPV-UHFFFAOYSA-N
Synonym 2-tributylstannyl thiazole, 2-tributylstannylthiazole, 2-tributylstannyl-1,3-thiazole, thiazole,2-tributylstannyl, tributylstannylthiazole, thiazole, 2-tributylstannyl, 2-tributylstannanyl-thiazole, 1,3-thiazol-2-yl tributylstannane, 2-tributylstannanyl-1,3-thiazole, 2-tri-n-butylstannyl thiazole
PubChem CID 2763259
IUPAC Name tributyl(1,3-thiazol-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=NC=CS1

Specifications

Density 1.19
Boiling Point 307°C to 309°C
Quantity 1 g
UN Number UN2788
Solubility Information Not miscible or difficult to mix.
Formula Weight 374.17
Percent Purity 96%
Chemical Name or Material 2-(Tri-n-butylstannyl)thiazole
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only