Learn More
2-(Tri-n-butylstannyl)pyrazine, 95%
CAS: 205371-27-3 | C16H30N2Sn | 369.14 g/mol
Supplier: Thermo Scientific Chemicals H5137003
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-(Tri-n-butylstannyl)pyrazine | |
| 1.171 | |
| >110°C (230°F) | |
| 1.516 | |
| 1 g | |
| 2-tributylstannyl pyrazine, 2-pyrazinyl tributyltin, 2-tributylstannylpyrazine, pyrazine, tributylstannyl, pyrazine,2-tributylstannyl, pyrazin-2-yl tributylstannane, tributylstannyl-pyrazine, 2-tri-n-butylstannyl pyrazine, tributyl-pyrazin-2-yl-stannane, tributylstannylpyrazine | |
| CCCC[Sn](CCCC)(CCCC)C1=NC=CN=C1 | |
| 369.14 | |
| 369.12 |
| 205371-27-3 | |
| 130°C (1 mmHg) | |
| C16H30N2Sn | |
| MFCD01319077 | |
| UN2788 | |
| OVBXTKIWZAHFAC-UHFFFAOYSA-N | |
| tributyl(pyrazin-2-yl)stannane | |
| 2737316 | |
| 95% |
Chemical Identifiers
| 205371-27-3 | |
| 369.14 | |
| OVBXTKIWZAHFAC-UHFFFAOYSA-N | |
| 2737316 | |
| CCCC[Sn](CCCC)(CCCC)C1=NC=CN=C1 |
| C16H30N2Sn | |
| MFCD01319077 | |
| 2-tributylstannyl pyrazine, 2-pyrazinyl tributyltin, 2-tributylstannylpyrazine, pyrazine, tributylstannyl, pyrazine,2-tributylstannyl, pyrazin-2-yl tributylstannane, tributylstannyl-pyrazine, 2-tri-n-butylstannyl pyrazine, tributyl-pyrazin-2-yl-stannane, tributylstannylpyrazine | |
| tributyl(pyrazin-2-yl)stannane |
Safety and Handling
GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only