missing translation for 'onlineSavingsMsg'
Learn More

2-(Tri-n-butylstannyl)pyrazine, 95%

Catalog No. p-7033985
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51370MD 250 mg
AAH5137003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAH51370MD Supplier Thermo Scientific Chemicals Supplier No. H51370MD
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 205371-27-3 | C16H30N2Sn | 369.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 205371-27-3
Molecular Formula C16H30N2Sn
Molecular Weight (g/mol) 369.14
MDL Number MFCD01319077
InChI Key OVBXTKIWZAHFAC-UHFFFAOYSA-N
Synonym 2-tributylstannyl pyrazine, 2-pyrazinyl tributyltin, 2-tributylstannylpyrazine, pyrazine, tributylstannyl, pyrazine,2-tributylstannyl, pyrazin-2-yl tributylstannane, tributylstannyl-pyrazine, 2-tri-n-butylstannyl pyrazine, tributyl-pyrazin-2-yl-stannane, tributylstannylpyrazine
PubChem CID 2737316
IUPAC Name tributyl(pyrazin-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=NC=CN=C1

Specifications

Density 1.171
Boiling Point 130°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.516
Quantity 250 mg
UN Number UN2788
Formula Weight 369.12
Percent Purity 95%
Chemical Name or Material 2-(Tri-n-butylstannyl)pyrazine
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only