missing translation for 'onlineSavingsMsg'
Learn More

2-Thioxanthine 80.0+%, TCI America™

Catalog No. T02251G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T02251G 1 g
T02255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T02251G Supplier TCI America Supplier No. T02251G

Chemical Identifiers

CAS 2487-40-3
Molecular Formula C5H4N4OS
Molecular Weight (g/mol) 168.174
MDL Number MFCD00031505
InChI Key XNHFAGRBSMMFKL-UHFFFAOYSA-N
Synonym 2-thioxanthine, 6-hydroxy-2-mercaptopurine, 2-mercapto-9h-purin-6-ol, 2-thioxanthene, 2-mercapto-6-hydroxypurine, 2-thio-6-oxypurine, 6-hydroxypurine-2-thiol, 2-thioxo-2,3-dihydro-1h-purin-6 7h-one, xanthine, 2-thio, 6h-purin-6-one, 1,2,3,7-tetrahydro-2-thioxo
PubChem CID 1268185
IUPAC Name 2-sulfanylidene-3,7-dihydropurin-6-one
SMILES C1=NC2=C(N1)C(=O)NC(=S)N2

Specifications

Quantity 1 g
Formula Weight 168.17
Percent Purity ≥80.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Thioxanthine
EINECSNumber (5)-1135
RTECSNumber UP0825000
TSCA Yes