accessibility menu, dialog, popup

UserName

ONHSNHNHN

CAS RN 2487-40-3

IUPAC Name: 2-sulfanylidene-2,3,6,7-tetrahydro-1H-purin-6-one
Synonyms: 2-sulfanylidene-3,7-dihydro-1H-purin-6-one
Molecular Weight (g/mol): 168.17
Molecular Formula: C5H4N4OS
InChi Key: XNHFAGRBSMMFKL-UHFFFAOYSA-N
SMILES: O=C1NC(=S)NC2=C1NC=N2
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)