missing translation for 'onlineSavingsMsg'
Learn More

2-Thioxanthine, 98+%

Catalog No. p-7025884
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2160603 1 g
AAB2160606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2160603 Supplier Thermo Scientific Chemicals Supplier No. B2160603

CAS: 2487-40-3 | C5H4N4OS | 168.174 g/mol

2-Thioxanthine used to inhibit MPO (Myeloperoxidase).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Thioxanthine used to inhibit MPO (Myeloperoxidase).

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 2487-40-3
Molecular Formula C5H4N4OS
Molecular Weight (g/mol) 168.174
MDL Number MFCD00031505
InChI Key XNHFAGRBSMMFKL-UHFFFAOYSA-N
Synonym 2-thioxanthine, 6-hydroxy-2-mercaptopurine, 2-mercapto-9h-purin-6-ol, 2-thioxanthene, 2-mercapto-6-hydroxypurine, 2-thio-6-oxypurine, 6-hydroxypurine-2-thiol, 2-thioxo-2,3-dihydro-1h-purin-6 7h-one, xanthine, 2-thio, 6h-purin-6-one, 1,2,3,7-tetrahydro-2-thioxo
PubChem CID 1268185
IUPAC Name 2-sulfanylidene-3,7-dihydropurin-6-one
SMILES C1=NC2=C(N1)C(=O)NC(=S)N2

Specifications

Melting Point >300°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 168.17
Percent Purity ≥98%
Chemical Name or Material 2-Thioxanthine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-636-7
RTECSNumber UP0825000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only