missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Thiophenemethanol 98.0+%, TCI America™
$192.00 - $2039.27
Chemical Identifiers
| CAS | 636-72-6 |
|---|---|
| Molecular Formula | C5H6OS |
| Molecular Weight (g/mol) | 114.16 |
| MDL Number | MFCD00005454 |
| InChI Key | ZPHGMBGIFODUMF-UHFFFAOYSA-N |
| Synonym | 2-thiophenemethanol, thiophene-2-methanol, 2-thienylmethanol, 2-hydroxymethylthiophene, 2-thenyl alcohol, 2-hydroxymethyl thiophene, thenyl alcohol, 2-thiophenecarbinol, thien-2-yl methanol, thiophen-2-yl methanol |
| PubChem CID | 69467 |
| IUPAC Name | (thiophen-2-yl)methanol |
| SMILES | OCC1=CC=CS1 |
Chemical Identifiers
| 636-72-6 | |
| 114.16 | |
| ZPHGMBGIFODUMF-UHFFFAOYSA-N | |
| 69467 | |
| OCC1=CC=CS1 |
| C5H6OS | |
| MFCD00005454 | |
| 2-thiophenemethanol, thiophene-2-methanol, 2-thienylmethanol, 2-hydroxymethylthiophene, 2-thenyl alcohol, 2-hydroxymethyl thiophene, thenyl alcohol, 2-thiophenecarbinol, thien-2-yl methanol, thiophen-2-yl methanol | |
| (thiophen-2-yl)methanol |
Specifications
| 636-72-6 | |
| 207°C | |
| MFCD00005454 | |
| 2-thiophenemethanol, thiophene-2-methanol, 2-thienylmethanol, 2-hydroxymethylthiophene, 2-thenyl alcohol, 2-hydroxymethyl thiophene, thenyl alcohol, 2-thiophenecarbinol, thien-2-yl methanol, thiophen-2-yl methanol | |
| OCC1=CC=CS1 | |
| 114.16 | |
| 114.16 | |
| Liquid |
| Yellow | |
| C5H6OS | |
| 25 g | |
| ZPHGMBGIFODUMF-UHFFFAOYSA-N | |
| (thiophen-2-yl)methanol | |
| 69467 | |
| ≥98.0% (GC) | |
| 2-Thiophenemethanol |
Safety and Handling
TSCA : No