missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenemethanol 98.0+%, TCI America™

Catalog No. T111225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T111225G 25 g
T1112100G 100 g
T1112500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T111225G Supplier TCI America Supplier No. T111225G

Chemical Identifiers

CAS 636-72-6
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.16
MDL Number MFCD00005454
InChI Key ZPHGMBGIFODUMF-UHFFFAOYSA-N
Synonym 2-thiophenemethanol, thiophene-2-methanol, 2-thienylmethanol, 2-hydroxymethylthiophene, 2-thenyl alcohol, 2-hydroxymethyl thiophene, thenyl alcohol, 2-thiophenecarbinol, thien-2-yl methanol, thiophen-2-yl methanol
PubChem CID 69467
IUPAC Name (thiophen-2-yl)methanol
SMILES OCC1=CC=CS1

Specifications

Color Yellow
Boiling Point 207°C
Quantity 25 g
Formula Weight 114.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Thiophenemethanol