missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenecarboxaldehyde (stabilized with HQ) 98.0+%, TCI America™

Catalog No. T072525ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T072525ML 25 mL
T0725100ML 100 mL
T0725500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T072525ML Supplier TCI America Supplier No. T072525ML

Chemical Identifiers

CAS 98-03-3
Molecular Formula C5H4OS
Molecular Weight (g/mol) 112.15
MDL Number MFCD00005429
InChI Key CNUDBTRUORMMPA-UHFFFAOYSA-N
Synonym 2-thiophenecarboxaldehyde, 2-thenaldehyde, thiophene-2-aldehyde, 2-formylthiophene, 2-thienylaldehyde, 2-thiophenealdehyde, thiophene-2-carboxaldehyde, 2-thienylcarboxaldehyde, 2-thienaldehyde, alpha-formylthiophene
PubChem CID 7364
ChEBI CHEBI:87301
IUPAC Name thiophene-2-carbaldehyde
SMILES O=CC1=CC=CS1

Specifications

Color Yellow
Boiling Point 198°C
Quantity 25 mL
Formula Weight 112.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Thiophenecarboxaldehyde (stabilized with HQ)
EINECSNumber (5)-0073
RTECSNumber XM8135000
TSCA Yes
Recommended Storage Refrigerator