missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Thiophenecarboxaldehyde, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Color | Yellow to Brown |
| Density | 1.2000g/mL |
| Boiling Point | 198.0°C |
| Flash Point | 77°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.5890 to 1.5920 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Warning |
| missing translation for 'healthHazard2' | GHS H Statement: May cause an allergic skin reaction. Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. |
| missing translation for 'healthHazard3' | GHS P Statement: Wear eye protection/face protection. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 202-629-8 |
| RTECSNumber | XM8135000 |
| TSCA | TSCA |
| Recommended Storage | Product may darken on standing |
RUO – Research Use Only