missing translation for 'onlineSavingsMsg'
Learn More

2-Thenoyl Chloride 99.0+%, TCI America™

Catalog No. T0177500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0177500G 500 g
T017725G 25 g
T0177100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T0177500G Supplier TCI America Supplier No. T0177500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5271-67-0
Molecular Formula C5H3ClOS
Molecular Weight (g/mol) 146.59
MDL Number MFCD00005428
InChI Key QIQITDHWZYEEPA-UHFFFAOYSA-N
Synonym 2-thiophenecarbonyl chloride, 2-thenoyl chloride, 2-thienylcarbonyl chloride, 2-thiophenecarbonylchloride, thiophenecarbonyl chloride, 2-thiophene carbonyl chloride, thiophene-2-carboxylic acid chloride, thenoyl chloride, thienoyl chloride, 2-thenoylchloride
PubChem CID 78928
IUPAC Name thiophene-2-carbonyl chloride
SMILES ClC(=O)C1=CC=CS1

Specifications

Boiling Point 190°C
Quantity 500 g
UN Number 3265
Formula Weight 146.59
Percent Purity ≥99.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Thenoyl Chloride