missing translation for 'onlineSavingsMsg'
Learn More

2-(Tetrahydropyran-4-yloxy)benzoic acid, 97%, Thermo Scientific™

Catalog No. p-3830063
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC52201CB 250mg Amber Glass Bottle
CC52201DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC52201CB Supplier Thermo Scientific Chemicals Supplier No. CC52201CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 898289-29-7
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD09025869
InChI Key AVRDARROMNUMER-UHFFFAOYSA-N
Synonym 2-oxan-4-yloxy benzoic acid, 2-tetrahydropyran-4-yloxy benzoic acid, 2-oxan-4-yl oxy benzoic acid, 2-tetrahydro-2h-pyran-4-yl oxy benzoic acid, benzoic acid, 2-tetrahydro-2h-pyran-4-yl oxy, benzoic acid,2-tetrahydro-2h-pyran-4-yl oxy, 2-2h-3,4,5,6-tetrahydropyran-4-yloxy benzoic acid
PubChem CID 21754839
IUPAC Name 2-(oxan-4-yloxy)benzoic acid
SMILES OC(=O)C1=CC=CC=C1OC1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 222.24
Percent Purity 97%
Chemical Name or Material 2-(Tetrahydropyran-4-yloxy)benzoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.