missing translation for 'onlineSavingsMsg'
Learn More

2-(Tetrahydropyran-4-yloxy)benzoic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC52201DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC52201DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

898289-29-7
222.24
AVRDARROMNUMER-UHFFFAOYSA-N
21754839
OC(=O)C1=CC=CC=C1OC1CCOCC1
C12H14O4
MFCD09025869
2-oxan-4-yloxy benzoic acid, 2-tetrahydropyran-4-yloxy benzoic acid, 2-oxan-4-yl oxy benzoic acid, 2-tetrahydro-2h-pyran-4-yl oxy benzoic acid, benzoic acid, 2-tetrahydro-2h-pyran-4-yl oxy, benzoic acid,2-tetrahydro-2h-pyran-4-yl oxy, 2-2h-3,4,5,6-tetrahydropyran-4-yloxy benzoic acid
2-(oxan-4-yloxy)benzoic acid

Specifications

2-(Tetrahydropyran-4-yloxy)benzoic acid
96.5
97%
C12H14O4
MFCD09025869
AVRDARROMNUMER-UHFFFAOYSA-N
2-(oxan-4-yloxy)benzoic acid
21754839
97%
898289-29-7
100.0
Amber Glass Bottle
1g
2-oxan-4-yloxy benzoic acid, 2-tetrahydropyran-4-yloxy benzoic acid, 2-oxan-4-yl oxy benzoic acid, 2-tetrahydro-2h-pyran-4-yl oxy benzoic acid, benzoic acid, 2-tetrahydro-2h-pyran-4-yl oxy, benzoic acid,2-tetrahydro-2h-pyran-4-yl oxy, 2-2h-3,4,5,6-tetrahydropyran-4-yloxy benzoic acid
OC(=O)C1=CC=CC=C1OC1CCOCC1
222.24
222.24
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents