missing translation for 'onlineSavingsMsg'
Learn More

2-Tetrahydropyran-4-ylethanol, 97%, Thermo Scientific™

$163.66 - $163.66

Chemical Identifiers

CAS 4677-18-3
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00129068
InChI Key XZXZZACRGBBWTQ-UHFFFAOYSA-N
Synonym 2-tetrahydro-2h-pyran-4-yl ethanol, 2-oxan-4-yl ethan-1-ol, 2-tetrahydro-pyran-4-yl-ethanol, 2-tetrahydropyran-4-ylethanol, 2-oxan-4-yl ethanol, 4-2-hydroxyethyl oxane, 4-hydroxyethyl tetrahydropyran, 2h-pyran-4-ethanol, tetrahydro, 2-tetrahydro-2h-pyran-4-yl ethan-1-ol, 2-tetrahydro-2h-pyran-4-ylethanol
PubChem CID 17750944
SMILES OCCC1CCOCC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC67409CB
SDS View Documents
Thermo Scientific Chemicals
CC67409CB
250 mg
Each for $163.66
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4677-18-3
130.19
XZXZZACRGBBWTQ-UHFFFAOYSA-N
17750944
C7H14O2
MFCD00129068
2-tetrahydro-2h-pyran-4-yl ethanol, 2-oxan-4-yl ethan-1-ol, 2-tetrahydro-pyran-4-yl-ethanol, 2-tetrahydropyran-4-ylethanol, 2-oxan-4-yl ethanol, 4-2-hydroxyethyl oxane, 4-hydroxyethyl tetrahydropyran, 2h-pyran-4-ethanol, tetrahydro, 2-tetrahydro-2h-pyran-4-yl ethan-1-ol, 2-tetrahydro-2h-pyran-4-ylethanol
OCCC1CCOCC1

Specifications

4677-18-3
100.0
Amber Glass Bottle
MFCD00129068
2-tetrahydro-2h-pyran-4-yl ethanol, 2-oxan-4-yl ethan-1-ol, 2-tetrahydro-pyran-4-yl-ethanol, 2-tetrahydropyran-4-ylethanol, 2-oxan-4-yl ethanol, 4-2-hydroxyethyl oxane, 4-hydroxyethyl tetrahydropyran, 2h-pyran-4-ethanol, tetrahydro, 2-tetrahydro-2h-pyran-4-yl ethan-1-ol, 2-tetrahydro-2h-pyran-4-ylethanol
OCCC1CCOCC1
130.19
130.19
2-Tetrahydropyran-4-ylethanol
96.5
97%
C7H14O2
250 mg
XZXZZACRGBBWTQ-UHFFFAOYSA-N
2-(oxan-4-yl)ethanol
17750944
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'