missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butyltoluene 99.0+%, TCI America™

Catalog No. B09661ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09661ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B09661ML Supplier TCI America Supplier No. B09661ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1074-92-6
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00059209
InChI Key AXHVNJGQOJFMHT-UHFFFAOYSA-N
Synonym 2-tert-butyltoluene, 1-tert-butyl-2-methylbenzene, toluene, o-tert-butyl, benzene, 1-1,1-dimethylethyl-2-methyl, benzene, 1,1-dimethylethyl methyl, o-tert-butyltoluene, 1-methyl-2-tert-butylbenzene, benzene,1-1,1-dimethylethyl-2-methyl, t-butyltoluene, o-t-butyltoluene
PubChem CID 33712
IUPAC Name 1-tert-butyl-2-methylbenzene
SMILES CC1=CC=CC=C1C(C)(C)C

Specifications

Color Yellow
Boiling Point 200°C
Quantity 1 mL
UN Number 2667
Formula Weight 148.25
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-tert-Butyltoluene
EINECSNumber (3)-0015
TSCA No