missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butyltoluene, 99%

Catalog No. AAH33036MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33036MD 250 mg
AAH3303603 1 g
AAH3303606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH33036MD Supplier Thermo Scientific Chemicals Supplier No. H33036MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1074-92-6 | C11H16 | 148.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1074-92-6
Molecular Formula C11H16
Molecular Weight (g/mol) 148.249
MDL Number MFCD00059209
InChI Key AXHVNJGQOJFMHT-UHFFFAOYSA-N
PubChem CID 33712
IUPAC Name 1-tert-butyl-2-methylbenzene
SMILES CC1=CC=CC=C1C(C)(C)C

Specifications

Density 0.89
Refractive Index 1.5075
Quantity 250 mg
UN Number UN2667
Synonym 2-tert-butyltoluene, 1-tert-butyl-2-methylbenzene, toluene, o-tert-butyl, benzene, 1-1,1-dimethylethyl-2-methyl, benzene, 1,1-dimethylethyl methyl, o-tert-butyltoluene, 1-methyl-2-tert-butylbenzene, benzene,1-1,1-dimethylethyl-2-methyl, t-butyltoluene, o-t-butyltoluene
Formula Weight 148.25
Percent Purity 99%
Chemical Name or Material 2-tert-Butyltoluene
Hazard Category H227-H302+H332-H319-H335
Hazard Statement GHS H Statement
H331-H227-H302-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BUTYLTOLUENES
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only