missing translation for 'onlineSavingsMsg'
Learn More

2-(tert-Butyldimethylsilyl)-N,N-dimethyl-1H-imidazole-1-sulfonamide 97.0+%, TCI America™

Catalog No. B56581G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56581G 1 g
B56585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B56581G Supplier TCI America Supplier No. B56581G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 129378-52-5
Molecular Formula C11H23N3O2SSi
Molecular Weight (g/mol) 289.469
MDL Number MFCD12963638
InChI Key TZBRCDMGXKRTCN-UHFFFAOYSA-N
Synonym 1-(Dimethylsulfamoyl)-2-(tert-butyldimethylsilyl)-1H-imidazole
PubChem CID 6420209
IUPAC Name 2-[tert-butyl(dimethyl)silyl]-N,N-dimethylimidazole-1-sulfonamide
SMILES CC(C)(C)[Si](C)(C)C1=NC=CN1S(=O)(=O)N(C)C

Specifications

Melting Point 64°C
Quantity 1 g
Formula Weight 289.47
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(tert-Butyldimethylsilyl)-N,N-dimethyl-1H-imidazole-1-sulfonamide