accessibility menu, dialog, popup

UserName

CAS RN 129378-52-5

IUPAC Name: 2-(tert-butyldimethylsilyl)-N,N-dimethyl-1H-imidazole-1-sulfonamide
Synonyms: 2-(tert-butyldimethylsilyl)-N,N-dimethylimidazole-1-sulfonamide
Molecular Weight (g/mol): 289.47
Molecular Formula: C11H23N3O2SSi
InChi Key: TZBRCDMGXKRTCN-UHFFFAOYSA-N
SMILES: CN(C)S(=O)(=O)N1C=CN=C1[Si](C)(C)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results