missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butylanthraquinone 98.0+%, TCI America™

Catalog No. B0816500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0816500G 500 g
B081625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0816500G Supplier TCI America Supplier No. B0816500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84-47-9
Molecular Formula C18H16O2
Molecular Weight (g/mol) 264.32
MDL Number MFCD00001232
InChI Key YTPSFXZMJKMUJE-UHFFFAOYSA-N
Synonym 2-tert-butylanthraquinone, 9,10-anthracenedione, 2-1,1-dimethylethyl, 2-tert-butyl-9,10-anthraquinone, 2-tert-butyl anthracene-9,10-dione, anthraquinone, 2-tert-butyl, beta-tert-butylanthraquinone, 2-t-butylanthraquinone, .beta.-tert-butylanthraquinone, acmc-209puu, 2-tert-butyl-anthrachinon
PubChem CID 66532
IUPAC Name 2-tert-butyl-9,10-dihydroanthracene-9,10-dione
SMILES CC(C)(C)C1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1

Specifications

Melting Point 104°C
Color Yellow
Quantity 500 g
UN Number 3077
Formula Weight 264.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-tert-Butylanthraquinone
RTECSNumber CB0487500
TSCA Yes