accessibility menu, dialog, popup

UserName

CAS RN 84-47-9

IUPAC Name: 2-tert-butyl-9,10-dihydroanthracene-9,10-dione
Synonyms: 2-tert-butylanthracene-9,10-dione
Molecular Weight (g/mol): 264.32
Molecular Formula: C18H16O2
InChi Key: YTPSFXZMJKMUJE-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results