missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butylanthracene 98.0+%, TCI America™

Catalog No. B35411G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35411G 1 g
B35415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35411G Supplier TCI America Supplier No. B35411G
Only null left

Chemical Identifiers

CAS 18801-00-8
Molecular Formula C18H18
Molecular Weight (g/mol) 234.342
MDL Number MFCD00003581
InChI Key WBPXZSIKOVBSAS-UHFFFAOYSA-N
PubChem CID 87800
IUPAC Name 2-tert-butylanthracene
SMILES CC(C)(C)C1=CC2=CC3=CC=CC=C3C=C2C=C1

Specifications

Melting Point 148°C
Color White-Yellow
Quantity 1 g
Synonym 2-tert-butyl anthracene, anthracene, 2-1,1-dimethylethyl, anthracene,2-1,1-dimethylethyl, 2-tertbutylanthracene, 2-t-butyl anthracene, acmc-209eqy
Formula Weight 234.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-tert-Butylanthracene