missing translation for 'onlineSavingsMsg'
Learn More

2-(tert-Butylamino)-4-(cyclopropylamino)-6-(methylthio)-1,3,5-triazine 98.0+%, TCI America™

Catalog No. I08425G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08425G 5 g
I084225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08425G Supplier TCI America Supplier No. I08425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 28159-98-0
Molecular Formula C11H19N5S
Molecular Weight (g/mol) 253.368
MDL Number MFCD01863779
InChI Key HDHLIWCXDDZUFH-UHFFFAOYSA-N
Synonym Cybutryne
PubChem CID 91590
ChEBI CHEBI:5962
IUPAC Name 2-N-tert-butyl-4-N-cyclopropyl-6-methylsulfanyl-1,3,5-triazine-2,4-diamine
SMILES CC(C)(C)NC1=NC(=NC(=N1)NC2CC2)SC

Specifications

Melting Point 128°C
Color White
Quantity 5 g
UN Number 3077
Formula Weight 253.37
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(tert-Butylamino)-4-(cyclopropylamino)-6-(methylthio)-1,3,5-triazine
EINECSNumber (5)-6110
RTECSNumber XY5850675
TSCA No