accessibility menu, dialog, popup

UserName

CH3SNHNNNHH3CCH3H3CN

CAS RN 28159-98-0

IUPAC Name: N2-tert-butyl-N4-cyclopropyl-6-(methylsulfanyl)-1,3,5-triazine-2,4-diamine
Synonyms: irgarol 1051 cybutryne
Molecular Weight (g/mol): 253.37
Molecular Formula: C11H19N5S
InChi Key: HDHLIWCXDDZUFH-UHFFFAOYSA-N
SMILES: CSC1=NC(NC2CC2)=NC(NC(C)(C)C)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results