missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butylamino-1-(4-hydroxy-3-methylphenyl)ethanol sulfate, MilliporeSigma™ Supelco™

Catalog No. 111002265
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002265 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002265 Supplier Merck Emd Millipore Supplier No. BP192
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Specifications

CAS 1956308-05-6
Linear Formula C13H23 NO6 S
Type Pharmacopeia and Metrological Institutes Standards
Quantity 25 mg
Formula Weight 321.39
Grade Pharmaceutical Primary Standard