missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butyl-trans-vinylboronic acid, 97%

CAS: 86595-37-1 | C6H13BO2 | 127.978 g/mol

Supplier:  Thermo Scientific Chemicals H5262703

 View more versions of this product

Catalog No. AAH5262703


Only null left
Explore available promotions

Description

Description

2-tert-Butyl-trans-vinylboronic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-tert-Butyl-trans-vinylboronic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2-tert-Butyl-trans-vinylboronic acid
78°C to 80°C
MFCD01074624
3,3-dimethyl-1-butenylboronic acid, e-3,3-dimethylbut-1-en-1-yl boronic acid, 2-t-butyl-e-vinylboronic acid, 2-tert-butyl-e-vinylboronic acid, e-3,3-dimethylbut-1-enyl boronic acid, 1-butenyl-3,3-dimethyboronic acid, 1e-3,3-dimethylbut-1-en-1-ylboronic acid, boronic acid, b-3,3-dimethyl-1-buten-1-yl, 3,3-dimethylbut-1-enylboronic acid
CTIWKIMYFQSVBZ-SNAWJCMRSA-N
[(E)-3,3-dimethylbut-1-enyl]boronic acid
5706775
97%
86595-37-1
C6H13BO2
1 g
Slightly soluble in water.
B(C=CC(C)(C)C)(O)O
127.978
127.98

Chemical Identifiers

86595-37-1
127.978
CTIWKIMYFQSVBZ-SNAWJCMRSA-N
5706775
B(C=CC(C)(C)C)(O)O
C6H13BO2
MFCD01074624
3,3-dimethyl-1-butenylboronic acid, e-3,3-dimethylbut-1-en-1-yl boronic acid, 2-t-butyl-e-vinylboronic acid, 2-tert-butyl-e-vinylboronic acid, e-3,3-dimethylbut-1-enyl boronic acid, 1-butenyl-3,3-dimethyboronic acid, 1e-3,3-dimethylbut-1-en-1-ylboronic acid, boronic acid, b-3,3-dimethyl-1-buten-1-yl, 3,3-dimethylbut-1-enylboronic acid
[(E)-3,3-dimethylbut-1-enyl]boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only