missing translation for 'onlineSavingsMsg'
Learn More

2-Sulfobenzaldehyde Sodium Salt 98.0+%, TCI America™

Catalog No. s012225g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
S012225G 25 g
S0122500G 500 g
2 options

Catalog No. S012225G

Supplier: TCI America S012225G

Only null left

Chemical Identifiers

CAS 1008-72-6
Molecular Formula C7H5NaO4S
Molecular Weight (g/mol) 208.16
MDL Number MFCD00007478
InChI Key ADPUQRRLAAPXGT-UHFFFAOYSA-M
Synonym sodium 2-formylbenzenesulfonate, 2-formylbenzenesulfonic acid sodium salt, sodium o-formylbenzenesulfonate, benzenesulfonic acid, 2-formyl-, sodium salt, unii-lh8xh59068, 2-sulfobenzaldehyde sodium salt, sodium benzaldehyde-2-sulfonate, benzaldehyde-2-sulfonic acid sodium salt, o-formylbenzenesulfonic acid sodium salt, sodium 2-formylbenzene-1-sulfonate
PubChem CID 3794540
IUPAC Name sodium 2-formylbenzene-1-sulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=CC=C1C=O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 208.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Sulfobenzaldehyde Sodium Salt
EINECSNumber (3)-2107
TSCA Yes