accessibility menu, dialog, popup

UserName

CAS RN 1008-72-6

IUPAC Name: sodium 2-formylbenzene-1-sulfonate
Synonyms: sodium 2-formylbenzenesulfonate
Molecular Weight (g/mol): 208.16
Molecular Formula: C7H5NaO4S
InChi Key: ADPUQRRLAAPXGT-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=CC=C1C=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)