missing translation for 'onlineSavingsMsg'
Learn More

2-Sulfobenzaldehyde Sodium Salt 98.0+%, TCI America™

Catalog No. p-7135823
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S012225G 25 g
S0122500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S012225G Supplier TCI America Supplier No. S012225G
Only null left

Chemical Identifiers

CAS 1008-72-6
Molecular Formula C7H5NaO4S
Molecular Weight (g/mol) 208.16
MDL Number MFCD00007478
InChI Key ADPUQRRLAAPXGT-UHFFFAOYSA-M
Synonym sodium 2-formylbenzenesulfonate, 2-formylbenzenesulfonic acid sodium salt, sodium o-formylbenzenesulfonate, benzenesulfonic acid, 2-formyl-, sodium salt, unii-lh8xh59068, 2-sulfobenzaldehyde sodium salt, sodium benzaldehyde-2-sulfonate, benzaldehyde-2-sulfonic acid sodium salt, o-formylbenzenesulfonic acid sodium salt, sodium 2-formylbenzene-1-sulfonate
PubChem CID 3794540
IUPAC Name sodium 2-formylbenzene-1-sulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=CC=C1C=O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 208.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Sulfobenzaldehyde Sodium Salt
EINECSNumber (3)-2107
TSCA Yes