missing translation for 'onlineSavingsMsg'
Learn More

2-Stearoylglycerol, ≥95% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014926
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014926 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014926 Supplier Merck Emd Millipore Supplier No. 3648425MG
Food and Beverages

Specifications

CAS 621-61-4
Physical Form Neat
Linear Formula C21H42O4
MDL Number MFCD25974054
Synonym beta-Monostearin; 2-Monostearin; 2-Monostearoylglycerol; 2-Octadecanoylglycerol; Glycerol beta-monostearate; Octadecanoic acid 2-hydroxy-1-(hydroxymethyl)ethyl ester
Type Analytical Standard
Quantity 25 mg
Formula Weight 358.56
Percent Purity ≥95% (GC)
Grade Analytical Standard