missing translation for 'onlineSavingsMsg'
Learn More

2-sec-Butylphenol 97.0+%, TCI America™

Catalog No. B0732500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0732500G 500 g
B073225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0732500G Supplier TCI America Supplier No. B0732500G

Chemical Identifiers

CAS 89-72-5
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00002225
InChI Key NGFPWHGISWUQOI-UHFFFAOYSA-N
Synonym 2-sec-butylphenol, o-sec-butylphenol, 2-1-methylpropyl phenol, 2-2-butyl phenol, phenol, 2-1-methylpropyl, 2-butan-2-yl phenol, phenol, o-sec-butyl, 2-sec.butylfenol, isopropyl o-cresol, 2-sec.butylfenol czech
PubChem CID 6984
ChEBI CHEBI:34303
IUPAC Name 2-butan-2-ylphenol
SMILES CCC(C)C1=CC=CC=C1O

Specifications

Color Yellow
Boiling Point 228°C
Quantity 500 g
UN Number 3145
Formula Weight 150.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-sec-Butylphenol
EINECSNumber (3)-0503
RTECSNumber SJ8920000
TSCA Yes