missing translation for 'onlineSavingsMsg'
Learn More

2-sec-Butylphenol, 98%

Catalog No. p-7024176
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1846130 250 g
AAA184610B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1846130 Supplier Thermo Scientific Chemicals Supplier No. A1846130
Only null left

CAS: 89-72-5 | C10H14O | 150.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89-72-5
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00002225
InChI Key NGFPWHGISWUQOI-UHFFFAOYSA-N
Synonym 2-sec-butylphenol, o-sec-butylphenol, 2-1-methylpropyl phenol, 2-2-butyl phenol, phenol, 2-1-methylpropyl, 2-butan-2-yl phenol, phenol, o-sec-butyl, 2-sec.butylfenol, isopropyl o-cresol, 2-sec.butylfenol czech
PubChem CID 6984
ChEBI CHEBI:34303
IUPAC Name 2-butan-2-ylphenol
SMILES CCC(C)C1=CC=CC=C1O

Specifications

Melting Point 10°C to 12°C
Density 0.982
Boiling Point 226°C to 228°C
Flash Point 112°C (233°F)
Odor Phenol-like
Refractive Index 1.522
Quantity 250 g
UN Number UN3145
Beilstein 1210026
Formula Weight 150.22
Percent Purity 98%
Chemical Name or Material 2-sec-Butylphenol
Show More Show Less
Hazard Category H302-H314-H331
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 201-933-8
RTECSNumber SJ8920000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only