missing translation for 'onlineSavingsMsg'
Learn More

2-[(RS)-(4-Methylphenyl)phenylmethoxy]-N,N-dimethylethanamine hydrochloride, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014147
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014147 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014147 Supplier Merck Emd Millipore Supplier No. 6222650MG
Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 4024-34-4
Linear Formula C18H23 NO · HCl
MDL Number MFCD00550569
Synonym Diphenhydramine impurity B (PhEur); 2-[(4-Methyl-alpha-phenylbenzyl)oxy]ethyl(dimethyl)ammonium chloride; 4-Methyldiphenhydramine hydrochloride; N,N-Dimethyl-2-[(4-methylphenyl)phenylmethoxy]ethanamin hydrochloride; Neobenodine
Type Pharmaceutical Impurity Standard
Quantity 50 mg
Formula Weight 305.84
Percent Purity ≥98% (HPLC)