missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrrolidinone, 99%

Catalog No. AAA1533236
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1533236 500 g
AAA15332A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1533236 Supplier Thermo Scientific Chemicals Supplier No. A1533236
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 616-45-5 | C4H7NO | 85.106 g/mol

2-Pyrrolidinone is used as a derivative in pharmaceuticals such as cotinine, doxapram, piracetam, povidone and ethosuximide. It is used as a solvent and an intermediate in polymers, inkjet cartridges, tone and colorant products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pyrrolidinone is used as a derivative in pharmaceuticals such as cotinine, doxapram, piracetam, povidone and ethosuximide. It is used as a solvent and an intermediate in polymers, inkjet cartridges, tone and colorant products.

Solubility
Miscible with water and other organic solvents.

Notes
Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.

Chemical Identifiers

CAS 616-45-5
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.106
MDL Number MFCD00005270
InChI Key HNJBEVLQSNELDL-UHFFFAOYSA-N
Synonym 2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam
PubChem CID 12025
ChEBI CHEBI:36592
IUPAC Name pyrrolidin-2-one
SMILES C1CC(=O)NC1

Specifications

Melting Point 23°C to 25°C
Density 1.12
Boiling Point 245°C
Flash Point >110°C (230°F)
Refractive Index 1.4875
Quantity 500 g
Sensitivity Hygroscopic
Merck Index 14,8016
Solubility Information Miscible with water and other organic solvents.
Formula Weight 85.11
Percent Purity 99%
Chemical Name or Material 2-Pyrrolidinone
Show More Show Less
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 210-483-1
RTECSNumber UY5715000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only