missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrrolidin-2-ylpyridine, 95%, Thermo Scientific™

Chemical Identifiers

CAS 77790-61-5
Molecular Formula C9H12N2
Molecular Weight (g/mol) 148.21
InChI Key NDCZQFDBSPOUDF-UHFFFAOYSA-N
Synonym 2-pyrrolidin-2-yl pyridine, 2-2-pyrrolidinyl pyridine, 2-pyrrolidin-2-yl-pyridine, +--alpha-nornicotine, pyridine, 2-2-pyrrolidinyl, +--2-2-pyrrolidinyl pyridine, pyridine, 2-2-pyrrolidinyl-, +-, r-2-2-pyrrolidinyl-pyridine, alpha-nornicotine, 2,3'-pyridylpyrrolidine
PubChem CID 2771659
IUPAC Name 2-pyrrolidin-2-ylpyridine
SMILES C1CC(NC1)C2=CC=CC=N2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC446740010
SDS View Documents
Thermo Scientific Chemicals
446740010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC446740050
SDS View Documents
Thermo Scientific Chemicals
446740050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

77790-61-5
148.21
2-pyrrolidin-2-yl pyridine, 2-2-pyrrolidinyl pyridine, 2-pyrrolidin-2-yl-pyridine, +--alpha-nornicotine, pyridine, 2-2-pyrrolidinyl, +--2-2-pyrrolidinyl pyridine, pyridine, 2-2-pyrrolidinyl-, +-, r-2-2-pyrrolidinyl-pyridine, alpha-nornicotine, 2,3'-pyridylpyrrolidine
2-pyrrolidin-2-ylpyridine
C9H12N2
NDCZQFDBSPOUDF-UHFFFAOYSA-N
2771659
C1CC(NC1)C2=CC=CC=N2

Specifications

77790-61-5
94% min. (GC)
C9H12N2
2-pyrrolidin-2-yl pyridine, 2-2-pyrrolidinyl pyridine, 2-pyrrolidin-2-yl-pyridine, +--alpha-nornicotine, pyridine, 2-2-pyrrolidinyl, +--2-2-pyrrolidinyl pyridine, pyridine, 2-2-pyrrolidinyl-, +-, r-2-2-pyrrolidinyl-pyridine, alpha-nornicotine, 2,3'-pyridylpyrrolidine
C1CC(NC1)C2=CC=CC=N2
148.21
148.21
2-Pyrrolidin-2-ylpyridine
82.0°C (0.8 mmHg)
Glass bottle
1g
NDCZQFDBSPOUDF-UHFFFAOYSA-N
2-pyrrolidin-2-ylpyridine
2771659
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only