accessibility menu, dialog, popup

UserName

HNN

CAS RN 77790-61-5

IUPAC Name: 2-(pyrrolidin-2-yl)pyridine
Synonyms: 2-(pyrrolidin-2-yl)pyridine
Molecular Weight (g/mol): 148.21
Molecular Formula: C9H12N2
InChi Key: NDCZQFDBSPOUDF-UHFFFAOYNA-N
SMILES: C1CNC(C1)C1=CC=CC=N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)