Learn More
2-Pyrimidinecarbonitrile, 98%
CAS: 14080-23-0 | C5H3N3 | 105.10 g/mol
$238.44 - $238.44
Chemical Identifiers
| CAS | 14080-23-0 |
|---|---|
| Molecular Formula | C5H3N3 |
| Molecular Weight (g/mol) | 105.10 |
| MDL Number | MFCD00160513 |
| InChI Key | IIHQNAXFIODVDU-UHFFFAOYSA-N |
| Synonym | 2-cyanopyrimidine, 2-pyrimidinecarbonitrile, 2-cyano-pyrimidine, 2-cyano pyrimidine, cyanopyrimidine, provastatin, pyrimidinecarbonitrile, zlchem 283, pubchem7042, pyrimidin-2-carbonitrile |
| PubChem CID | 2757979 |
| IUPAC Name | pyrimidine-2-carbonitrile |
| SMILES | N#CC1=NC=CC=N1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Qty | ||||
|
AC429820250
|
Thermo Scientific Chemicals
429820250 |
25 g | Glass bottle |
Each for $238.44
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 14080-23-0 | |
| 105.10 | |
| IIHQNAXFIODVDU-UHFFFAOYSA-N | |
| 2757979 | |
| N#CC1=NC=CC=N1 |
| C5H3N3 | |
| MFCD00160513 | |
| 2-cyanopyrimidine, 2-pyrimidinecarbonitrile, 2-cyano-pyrimidine, 2-cyano pyrimidine, cyanopyrimidine, provastatin, pyrimidinecarbonitrile, zlchem 283, pubchem7042, pyrimidin-2-carbonitrile | |
| pyrimidine-2-carbonitrile |
Specifications
| 14080-23-0 | |
| 100.0 | |
| White to Yellow | |
| Authentic | |
| Glass bottle | |
| MFCD00160513 | |
| 2-cyanopyrimidine, 2-pyrimidinecarbonitrile, 2-cyano-pyrimidine, 2-cyano pyrimidine, cyanopyrimidine, provastatin, pyrimidinecarbonitrile, zlchem 283, pubchem7042, pyrimidin-2-carbonitrile | |
| IIHQNAXFIODVDU-UHFFFAOYSA-N | |
| pyrimidine-2-carbonitrile | |
| 2757979 | |
| 98% | |
| 2-Pyrimidinecarbonitrile |
| 97.5 | |
| 37.0°C to 41.0°C | |
| 110°C | |
| 97.5% min. (GC) | |
| C5H3N3 | |
| 25 g | |
| Solubility in water: .. Other solubilities: soluble in methanol | |
| N#CC1=NC=CC=N1 | |
| 105.10 | |
| 105.1 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement
Toxic if swallowed.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or ras
GHS Signal Word: Danger
RUO – Research Use Only