missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridylacetic acid hydrochloride, 99%

Catalog No. p-189376
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC131950050 5 g Glass bottle
AC131950250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC131950050 Supplier Thermo Scientific Chemicals Supplier No. 131950050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16179-97-8 | C7H7NO2·HCl | 173.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16179-97-8
Molecular Formula C7H7NO2·HCl
Molecular Weight (g/mol) 173.6
MDL Number MFCD00012812
InChI Key MQVISALTZUNQSK-UHFFFAOYSA-N
Synonym 2-pyridylacetic acid hydrochloride, 2-pyridin-2-yl acetic acid hydrochloride, 2-pyridineacetic acid hydrochloride, 2-pyridineacetic acid, hydrochloride, pyridin-2-ylacetic acid hydrochloride, pyridylacetic acid hydrochloride, 2-pyridineacetic acid, hydrochloride 1:1, 2-pyridylacetic acid hydrochloide, pyridin-2-yl acetic acid hydrochloride, 2-2-pyridyl acetic acid hydrochloride
PubChem CID 85317
IUPAC Name 2-pyridin-2-ylacetic acid;hydrochloride
SMILES C1=CC=NC(=C1)CC(=O)O.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 135°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (C5H4N)CH2COOH·HCl
Quantity 5 g
Formula Weight 173.6
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Pyridylacetic acid hydrochloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 240-316-8
TSCA TSCA

RUO – Research Use Only