missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridinesulfonylacetonitrile, 98%

Catalog No. AAL0711403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0711403 1 g
AAL0711406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0711403 Supplier Thermo Scientific Chemicals Supplier No. L0711403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 170449-34-0 | C7H6N2O2S | 182.197 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 170449-34-0
Molecular Formula C7H6N2O2S
Molecular Weight (g/mol) 182.197
MDL Number MFCD00041443
InChI Key SAKACXZDAJXBCI-UHFFFAOYSA-N
Synonym 2-pyridinesulfonylacetonitrile, 2-pyridinesulphonylacetonitrile, 2-pyridin-2-ylsulfonyl acetonitrile, pyridine-2-sulfonylacetonitrile, 2-pyridine-2-sulfonyl acetonitrile, pyridin-2-ylsulfonyl acetonitrile, 2-2-pyridylsulfonyl ethanenitrile, maybridge1_004727, acmc-1c3jt, 2-pyridinesulfonlyacetonitrile
PubChem CID 519355
IUPAC Name 2-pyridin-2-ylsulfonylacetonitrile
SMILES C1=CC=NC(=C1)S(=O)(=O)CC#N

Specifications

Melting Point 77°C to 79°C
Quantity 1 g
UN Number UN3439
Formula Weight 182.21
Percent Purity 98%
Chemical Name or Material 2-Pyridinesulfonylacetonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only