Learn More
2-Pyridinesulfonic acid, 97%, Thermo Scientific™
$202.33
Chemical Identifiers
| CAS | 15103-48-7 |
|---|---|
| Molecular Formula | C5H5NO3S |
| Molecular Weight (g/mol) | 159.16 |
| MDL Number | MFCD00234080 |
| InChI Key | KZVLNAGYSAKYMG-UHFFFAOYSA-N |
| Synonym | 2-pyridinesulfonic acid, pyridinesulfonic acid, 2-pyridinesulfonic acid 8ci 9ci, alpha-pyridinesulfonic acid, sulfopyridin, pyridine-2-sulfonicacid, acmc-1bo1h, .alpha.-pyridinesulfonic acid, kzvlnagysakymg-uhfffaoysa |
| PubChem CID | 94849 |
| IUPAC Name | pyridine-2-sulfonic acid |
| SMILES | OS(=O)(=O)C1=CC=CC=N1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC455760050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 15103-48-7 | |
| 159.16 | |
| KZVLNAGYSAKYMG-UHFFFAOYSA-N | |
| 94849 | |
| OS(=O)(=O)C1=CC=CC=N1 |
| C5H5NO3S | |
| MFCD00234080 | |
| 2-pyridinesulfonic acid, pyridinesulfonic acid, 2-pyridinesulfonic acid 8ci 9ci, alpha-pyridinesulfonic acid, sulfopyridin, pyridine-2-sulfonicacid, acmc-1bo1h, .alpha.-pyridinesulfonic acid, kzvlnagysakymg-uhfffaoysa | |
| pyridine-2-sulfonic acid |
Specifications
| 15103-48-7 | |
| 96% min. (HPLC) | |
| MFCD00234080 | |
| Solubility in water: very soluble. | |
| OS(=O)(=O)C1=CC=CC=N1 | |
| 159.16 | |
| 159.16 | |
| 2-Pyridinesulfonic acid |
| Authentic | |
| C5H5NO3S | |
| 2-pyridinesulfonic acid, pyridinesulfonic acid, 2-pyridinesulfonic acid 8ci 9ci, alpha-pyridinesulfonic acid, sulfopyridin, pyridine-2-sulfonicacid, acmc-1bo1h, .alpha.-pyridinesulfonic acid, kzvlnagysakymg-uhfffaoysa | |
| KZVLNAGYSAKYMG-UHFFFAOYSA-N | |
| pyridine-2-sulfonic acid | |
| 94849 | |
| 97% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately
GHS Signal Word: Danger
RUO – Research Use Only